Learn More
Waters Corp Tube Spacer, 6 cc, 16 mm, for 75 mm Test Tubes
Catalog No. 502390658 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tube spacer is designed for use with the Otto SPEcialist Positive Pressure Manifold. It accommodates 6 cc tubes with a 16 mm diameter and is compatible with 75 mm test tubes.
  • Accommodates 6 cc tubes
  • Designed for 16 mm diameter tubes
  • Compatible with 75 mm test tubes
  • For use with the Otto SPEcialist Positive Pressure Manifold
Catalog No. 50-239-0658 Supplier Waters Corp Supplier No. 186009726
This tube spacer is designed for use with the Otto SPEcialist Positive Pressure Manifold. It accommodates 6 cc tubes with a 16 mm diameter and is compatible with 75 mm test tubes.
  • Accommodates 6 cc tubes
  • Designed for 16 mm diameter tubes
  • Compatible with 75 mm test tubes
  • For use with the Otto SPEcialist Positive Pressure Manifold