Learn More
Waters Corp Otto Cartridge, 6 mL, 13 mm, Flanged/Flangeless
Catalog No. 502392231 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Otto 6 mL flanged/flangeless cartridge is designed for use in positive pressure manifolds for efficient sample collection. Each cartridge is compatible with 13 mm collection tubes. This item is typically supplied as part of a Collection Tubes Startup Kit, which contains eighteen of these cartridges.
  • Flanged/flangeless design for versatility.
  • 6 mL capacity for sample collection.
  • Compatible with 13 mm collection tubes.
  • Suitable for positive pressure manifold systems.
Catalog No. 50-239-2231 Supplier Waters Corp Supplier No. 176004610
This Otto 6 mL flanged/flangeless cartridge is designed for use in positive pressure manifolds for efficient sample collection. Each cartridge is compatible with 13 mm collection tubes. This item is typically supplied as part of a Collection Tubes Startup Kit, which contains eighteen of these cartridges.
  • Flanged/flangeless design for versatility.
  • 6 mL capacity for sample collection.
  • Compatible with 13 mm collection tubes.
  • Suitable for positive pressure manifold systems.