Learn More
Waters Corp Oasis PRiME MCX Vac Cartridge, 6 cc, 150 mg, 30 μm
Catalog No. 502390501 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge is a 6 cc cartridge containing 150 mg of 30 μm particle size sorbent, supplied in boxes of 100. It is designed for targeted cleanup of basic compounds with pKa ≥ 4.5.
  • Vac cartridge format.
  • 6 cc volume capacity.
  • 150 mg sorbent amount.
  • 30 μm particle size.
  • Supplied in boxes of 100.
  • Highly efficient, orthogonal (reversed-phase and ion-exchange) solid-phase extraction product.
  • Uses a water-wettable strong cation-exchange/reversed-phase sorbent.
  • Removes up to 99% of phospholipids.
  • Does not require conditioning and equilibration steps.
  • Increased flow reproducibility and predictable processing time.
Catalog No. 50-239-0501 Supplier Waters Corp Supplier No. 186009722
This cartridge is a 6 cc cartridge containing 150 mg of 30 μm particle size sorbent, supplied in boxes of 100. It is designed for targeted cleanup of basic compounds with pKa ≥ 4.5.
  • Vac cartridge format.
  • 6 cc volume capacity.
  • 150 mg sorbent amount.
  • 30 μm particle size.
  • Supplied in boxes of 100.
  • Highly efficient, orthogonal (reversed-phase and ion-exchange) solid-phase extraction product.
  • Uses a water-wettable strong cation-exchange/reversed-phase sorbent.
  • Removes up to 99% of phospholipids.
  • Does not require conditioning and equilibration steps.
  • Increased flow reproducibility and predictable processing time.