Learn More
Waters Corp Waters Otto Drainless Waste Reservoir, Solid-Phase Extraction, 96 Well Plate
Catalog No. 502039147 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Otto Drainless Waste Reservoir is designed for solid-phase extraction (SPE) applications. It efficiently collects waste solutions generated during SPE pretreatment steps such as conditioning, equilibration, loading, and washing. This 96 well plate format reservoir is suitable for various sample pretreatment workflows.
  • Collects waste solutions generated during SPE pretreatment steps (conditioning, equilibration, loading, washing)
  • Usable after solvent disposal in each step
  • Solid-phase extraction product type
  • 96 well plate format
  • Suitable for sample pretreatment
  • One unit per package
Catalog No. 50-203-9147 Supplier Waters Corp Supplier No. 186009714
Add to Cart
Add to Cart
The Otto Drainless Waste Reservoir is designed for solid-phase extraction (SPE) applications. It efficiently collects waste solutions generated during SPE pretreatment steps such as conditioning, equilibration, loading, and washing. This 96 well plate format reservoir is suitable for various sample pretreatment workflows.
  • Collects waste solutions generated during SPE pretreatment steps (conditioning, equilibration, loading, washing)
  • Usable after solvent disposal in each step
  • Solid-phase extraction product type
  • 96 well plate format
  • Suitable for sample pretreatment
  • One unit per package