Learn More
Waters Corp Otto SPEcialist Positive Pressure Manifold, 96-Well Plate, SPE Cartridge, 4-Bed, 110 V
Catalog No. 502390568 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This compact and user-friendly device is designed for positive pressure extractions using 96-well plates and SPE cartridges, enhancing reproducibility and preventing cross-contamination.
  • Accommodates 1 cc, 3 cc, and 6 cc SPE cartridges, 96-well plates, and μElution plates.
  • Individual pressure and flow regulation for four manifold beds.
  • Universal design compatible with most SPE 96-well plates.
  • Inert flow path for all SPE solvents.
  • Controlled, reproducible flow rates.
  • Software for method development and saving protocols.
  • Vent to quickly release pressure.
  • Internal waste reservoir for easy emptying.
  • Compact design.
Catalog No. 50-239-0568 Supplier Waters Corp Supplier No. 725000682
Add to Cart
Add to Cart
Request a Quote
This compact and user-friendly device is designed for positive pressure extractions using 96-well plates and SPE cartridges, enhancing reproducibility and preventing cross-contamination.
  • Accommodates 1 cc, 3 cc, and 6 cc SPE cartridges, 96-well plates, and μElution plates.
  • Individual pressure and flow regulation for four manifold beds.
  • Universal design compatible with most SPE 96-well plates.
  • Inert flow path for all SPE solvents.
  • Controlled, reproducible flow rates.
  • Software for method development and saving protocols.
  • Vent to quickly release pressure.
  • Internal waste reservoir for easy emptying.
  • Compact design.