Learn More
Waters Corp Waters Corporation Waste Reservoir, with Drain, for Otto SPEcialist PPM
Catalog No. 502390653 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waste Reservoir with Drain is an optional accessory for the Otto SPEcialist Positive Pressure Manifold. It is designed to effectively collect solvent waste generated from the processing of SPE consumables.
  • Effectively collects solvent waste from SPE consumable processing.
  • Designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Requires a suitable waste container positioned below the bench top when in use.
Catalog No. 50-239-0653 Supplier Waters Corp Supplier No. 186009727
The Waste Reservoir with Drain is an optional accessory for the Otto SPEcialist Positive Pressure Manifold. It is designed to effectively collect solvent waste generated from the processing of SPE consumables.
  • Effectively collects solvent waste from SPE consumable processing.
  • Designed for use with the Otto SPEcialist Positive Pressure Manifold.
  • Requires a suitable waste container positioned below the bench top when in use.