Learn More
Aviva Systems Biology Corp OTUB2 Antibody - N-terminal region (ARP59281_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59281_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTUB2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human OTUB2. Alternate names: C14orf137, FLJ21916, MGC3102, OTB2, OTU2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-641