Learn More
Enzo Life Sciences Otubain-1 (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW06800100
Encompass_Preferred
Alternative name: OTUB1, Ubiquitin thioesterase OTUB1, Ubiquitin specific processing protease OTUB1, Deubiquitinating enzyme OTUB1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 50mM NaCl and 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2263