Learn More
Aviva Systems Biology Corp OTUD6A Antibody - middle region (ARP59277_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59277_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTUD6A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human OTUD6A. Alternate names: DUBA2, FLJ25831, HSHIN6ApplicationWB. Species reactivity: Human
Catalog No. 50-286-639