Learn More
Aviva Systems Biology Corp OTUD6B Antibody - middle region (ARP56809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTUD6B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OTUD6B. Alternate names: CGI-77, duba5, DUBA5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61996