Learn More
Apexbio Technology LLC OTX-015 202590-98-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063243
Encompass_Preferred
OTX-015 (CAS 202590-98-5) is a selective inhibitor of BRD2 BRD3 and BRD4 proteins within the BET (bromodomain and extra-terminal) family involved in transcriptional regulation By disrupting the interaction between these BRD proteins and acetylated histone H4 OTX-015 exhibits inhibitory activity demonstrated by IC50 values between 92 and 112 nM and in vitro EC50 ranging from 10 to 19 nM OTX-015 treatment reduces proliferation (GI50 60 200 nM) induces G1 arrest and promotes apoptosis in various human cancer cell models In xenograft mouse studies of BRD-NUT midline carcinoma orally administered OTX-015 significantly suppresses tumor growth showing potential utility for cancer research
Catalog No. 50-000-63243 Supplier Apexbio Technology LLC Supplier No. A36925.1
OTX-015 (CAS 202590-98-5) is a selective inhibitor of BRD2 BRD3 and BRD4 proteins within the BET (bromodomain and extra-terminal) family involved in transcriptional regulation By disrupting the interaction between these BRD proteins and acetylated histone H4 OTX-015 exhibits inhibitory activity demonstrated by IC50 values between 92 and 112 nM and in vitro EC50 ranging from 10 to 19 nM OTX-015 treatment reduces proliferation (GI50 60 200 nM) induces G1 arrest and promotes apoptosis in various human cancer cell models In xenograft mouse studies of BRD-NUT midline carcinoma orally administered OTX-015 significantly suppresses tumor growth showing potential utility for cancer research