Learn More
Selleck Chemical LLC OTX008-5mg
SDP
Catalog No. 502389958
Encompass_Preferred
OTX008 (Calixarene 0118, PTX008), a calixarene derivative, is a selective inhibitor of galectin-1 (Gal1) with anti-tumoural activity.
Catalog No. 50-238-9958 Supplier Selleck Chemical LLC Supplier No. S69495MG
OTX008 (Calixarene 0118, PTX008), a calixarene derivative, is a selective inhibitor of galectin-1 (Gal1) with anti-tumoural activity.