Learn More
Aviva Systems Biology Corp OTX1 Antibody - N-terminal region (P100957_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100957_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OTX1. Alternate names: FLJ38361, MGC15736ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62799