Learn More
Aviva Systems Biology Corp OTX2 Antibody - middle region (ARP32440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OTX2. Alternate names: MCOPS5, MGC45000, CPHD6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54589