Learn More
Aviva Systems Biology Corp OTX2 Antibody - N-terminal region (ARP32439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OTX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human OTX2. Alternate names: MCOPS5, MGC45000, CPHD6ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-52863