Learn More
TARGETMOL CHEMICALS INC OU749 10MG
SDP
Catalog No. 502580978
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OU749 is a (gamma)-glutamyl transferase (GGT) inhibitor with an intrinsic Ki of 17.6 uM. purity: 99%
Catalog No. 50-258-0978 Supplier TARGETMOL CHEMICALS INC Supplier No. T2212810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OU749 is a (gamma)-glutamyl transferase (GGT) inhibitor with an intrinsic Ki of 17.6 uM. purity: 99%