Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ouabagenin, MP Biomedicals™

Catalog No. ICN15600625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15600625 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15600625 Supplier MP Biomedicals, Inc Supplier No. 0215600625

Chemical Identifiers

CAS 508-52-1
Molecular Formula C23H34O8
Molecular Weight (g/mol) 438.517
InChI Key BXSABLKMKAINIU-QOHCMMFCSA-N
Synonym ouabagenin, g-strophanthidin, ouabain genin, 1beta,3beta,5,11alpha,14,19-hexahydroxy-5beta-card-20 22-enolide, 1-beta,3-beta,5,11-alpha,14,19-hexahydroxy-5-beta-card-20 22-enolide, 4-18-00-03554 beilstein handbook reference, 5-beta-card-20 22-enolide, 1-beta,3-beta,5,11-alpha,14,19-hexahydroxy, 1, a,3, a,5,11, a,14,19-hexahydroxy-5, a-card-20 22-enolide, 1 beta,3beta,5alpha,11alpha,14,19-hexa-hydroxy-5 beta,20 22-cardenolide
PubChem CID 12313812
ChEBI CHEBI:71016
IUPAC Name 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,3,5,11,14-pentahydroxy-10-(hydroxymethyl)-13-methyl-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
SMILES CC12CC(C3C(C1(CCC2C4=CC(=O)OC4)O)CCC5(C3(C(CC(C5)O)O)CO)O)O

Specifications

Color White
Quantity 25 mg
Formula Weight 438.5
Physical Form Powder
Chemical Name or Material Ouabagenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.