Learn More
Apexbio Technology LLC Ouabain,50mg CAS# 630-60-4
SDP
Catalog No. 502240098
Encompass_Preferred
Ouabain is a selective Na+/K+, -ATPase inhibitor, binds to α2 /α3 subunit with Ki of 41 nM/15 nM. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0098 Supplier Apexbio Technology LLC Supplier No. B227050
Ouabain is a selective Na+/K+, -ATPase inhibitor, binds to α2 /α3 subunit with Ki of 41 nM/15 nM. Other sizes are also available. Please inqury us for quote.