Learn More
Enzo Life Sciences Ouabain . octahydrate (100mg). CAS: 11018-89-6
SDP
Supplier:  Enzo Life Sciences ALX350066M100
Encompass_Preferred
A steroidal inhibitor of cardiac sodium pumps (Na+/K+ ATPase) which has been recognized as an endogenous adrenal hormone. In the CNS it can be used as a tool to induce in vivo excitotoxicity. Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens. Alternative name: Strophanthin G, 3-[(6-Deoxy-alpha-L-mannopyranosyl)oxy]-1,5,11alpha-14,19-penta-hydroxycard-20(22)-enolide. Purity: ≥98% (TLC). Solubility: Soluble in water (>25mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8716