Learn More
Apexbio Technology LLC Ouabain Octahydrate 11018-89-6 50mg
SDP
Catalog No. 5000094329
Encompass_Preferred
Ouabain Octahydrate (CAS 11018-89-6) is a cardiac glycoside that selectively inhibits the Na /K -ATPase enzyme thereby altering ion gradients across the plasma membrane Inhibition of this transporter impacts cellular homeostasis and has been shown to modulate the transcription of multidrug resistance (MDR) and multidrug resistance-associated protein (MRP) genes Ouabain Octahydrate is commonly utilized in biomedical research to study ion transport mechanisms investigate cellular resistance pathways and explore regulatory networks involved in drug efflux and sensitivity
Catalog No. 50-000-94329 Supplier Apexbio Technology LLC Supplier No. B343650
Ouabain Octahydrate (CAS 11018-89-6) is a cardiac glycoside that selectively inhibits the Na /K -ATPase enzyme thereby altering ion gradients across the plasma membrane Inhibition of this transporter impacts cellular homeostasis and has been shown to modulate the transcription of multidrug resistance (MDR) and multidrug resistance-associated protein (MRP) genes Ouabain Octahydrate is commonly utilized in biomedical research to study ion transport mechanisms investigate cellular resistance pathways and explore regulatory networks involved in drug efflux and sensitivity