Learn More
Cayman Chemical Ouabain (hydrate), Purity: ≥98%, CAS: 340169-01-9, 10 mg
SDP
Catalog No. 502953891
Encompass_Preferred
A cardiac glycoside with diverse biological activities; inhibits the Na+/K+-ATPase isoforms α1β1 and α2β1 and hERG (IC50s = 97, 90, and 15.2 nM, respectively); reduces the proliferation of PC3 and U937 cells (EC50s = 210 and 94 nM, respectively); inhibits the cytopathic effect of SARS-CoV-2 in infected Vero cells (IC50 = in vivo
Catalog No. 50-295-3891 Supplier Cayman Chemical Supplier No. 1431910 MG
A cardiac glycoside with diverse biological activities; inhibits the Na+/K+-ATPase isoforms α1β1 and α2β1 and hERG (IC50s = 97, 90, and 15.2 nM, respectively); reduces the proliferation of PC3 and U937 cells (EC50s = 210 and 94 nM, respectively); inhibits the cytopathic effect of SARS-CoV-2 in infected Vero cells (IC50 = in vivo