Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OUL35 (NSC39047) | 6336-34-1 | 99.8% | 256.26 g/mol | C14H12N2O3 | 25MG
SDP

Catalog No. 5000238713
Encompass_Preferred

OUL35 (NSC39047) is a selective small-molecule inhibitor of ARTD10 (PARP-10) with a reported IC50 of 329 nM. It is supplied as a high-purity research reagent intended for biochemical and cell-based studies, with solubility and storage guidance provided for in vitro and in vivo use.

  • Selective ARTD10 (PARP-10) inhibitor with IC50 of 329 nM.
  • High purity: 99.8% by supplier specification.
  • Molecular weight 256.26 g/mol; chemical formula C14H12N2O3.
  • Soluble in DMSO (≈83 mg/mL) with supplier formulation examples for in vivo use.
  • Available in multiple pack sizes and a 10 mM solution option for convenience.
  • Intended for biochemical and cellular research applications.

Catalog No. 50-002-38713 Supplier Medchemexpress LLC Supplier No. HY12351225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OUL35 (NSC39047) is a selective small-molecule inhibitor of ARTD10 (PARP-10) with a reported IC50 of 329 nM. It is supplied as a high-purity research reagent intended for biochemical and cell-based studies, with solubility and storage guidance provided for in vitro and in vivo use.

  • Selective ARTD10 (PARP-10) inhibitor with IC50 of 329 nM.
  • High purity: 99.8% by supplier specification.
  • Molecular weight 256.26 g/mol; chemical formula C14H12N2O3.
  • Soluble in DMSO (≈83 mg/mL) with supplier formulation examples for in vivo use.
  • Available in multiple pack sizes and a 10 mM solution option for convenience.
  • Intended for biochemical and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.