Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oul35 (NSC39047) | 6336-34-1 | 99.8% | 256.26 g/mol | C14H12N2O3 | 50MG
SDP

Catalog No. 5000235603
Encompass_Preferred

OUL35 (NSC39047) is a selective small-molecule inhibitor of ARTD10/PARP10 with an in vitro IC50 of 329 nM. It is supplied as a high-purity research compound for biochemical and cell-based assays probing ADP-ribosylation and PARP-related cellular processes.

  • Selective ARTD10/PARP10 inhibitor with reported IC50 of 329 nM.
  • High purity suitable for research applications (99.8%).
  • Molecular weight 256.26 g/mol; chemical formula C14H12N2O3.
  • Soluble in DMSO; store solid at 4°C and protect from light.
  • Available in small solid quantities and as DMSO solutions for assay setup.

Catalog No. 50-002-35603 Supplier Medchemexpress LLC Supplier No. HY12351250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OUL35 (NSC39047) is a selective small-molecule inhibitor of ARTD10/PARP10 with an in vitro IC50 of 329 nM. It is supplied as a high-purity research compound for biochemical and cell-based assays probing ADP-ribosylation and PARP-related cellular processes.

  • Selective ARTD10/PARP10 inhibitor with reported IC50 of 329 nM.
  • High purity suitable for research applications (99.8%).
  • Molecular weight 256.26 g/mol; chemical formula C14H12N2O3.
  • Soluble in DMSO; store solid at 4°C and protect from light.
  • Available in small solid quantities and as DMSO solutions for assay setup.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.