Learn More
Perkin Elmer US LLC General Purpose Optical Bench - MCT
Catalog No. 502016514
Encompass_Preferred
Our Frontier IR Systems, Spectrum 400 Series and Spectrum 100 Series instruments can be fitted with a General Purpose Optical Bench. The optical bench is supplied with a parabolic mirror assembly, an external MCT detector and a pre-amplifier module. The optical bench is bolted to the external accessory bracket, which is fitted in the handhold on the right of the instrument. The accessory uses the external beam port with an attached attenuator.
Catalog No. 50-201-6514 Supplier Perkin Elmer US LLC Supplier No. L1250425
Add to Cart
Add to Cart
Request a Quote
Our Frontier IR Systems, Spectrum 400 Series and Spectrum 100 Series instruments can be fitted with a General Purpose Optical Bench. The optical bench is supplied with a parabolic mirror assembly, an external MCT detector and a pre-amplifier module. The optical bench is bolted to the external accessory bracket, which is fitted in the handhold on the right of the instrument. The accessory uses the external beam port with an attached attenuator.