Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Outdoor Barricade Tape Roll - Woven Polypropylene, Horizontal Stripes, Magenta on Yellow

Catalog No. 19104315 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Magenta on Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-315 Magenta on Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-315 Supplier Brady™ Supplier No. 91173
Add to Cart
Add to Cart

Mark outdoor hazards with this woven barricade tape, made of B-903 polypropylene with ultraviolet inhibitors.

  • Made of B-903 Woven Polypropylene from Brady™
  • Contains special ultraviolet inhibitors for permanent outdoor marking
  • Visual barricading provides fast on-the-spot warning of potential danger
  • Woven barricade tape with black & yellow horizontal warning stripes

Specifications

Color Magenta on Yellow
Length (English) 150 ft.
Displayed Text Blank
Material Polypropylene
Type Barricade Tape
Certifications/Compliance OSHA 1910.144
Packaging Quantity 150 ft./Roll
Width (English) 0.75 in.
Width (Metric) 148.59 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.