Learn More
Grainger Outdoor Speaker Strobe Marked Fire White
Catalog No. 502767509
Encompass_Preferred
The Grainger 16X452 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end bell. This motor can be used for a wide variety of industrial and commercial applications. With its powerful 1/2 HP motor, it provides reliable performance. The open drip proof design allows it to be used in many environments. This electric motor would make a great addition for powering pumps, fans, conveyors, compressors, machine tools, and more.
Catalog No. 50-276-7509 Supplier Grainger Supplier No. 16X452
The Grainger 16X452 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end bell. This motor can be used for a wide variety of industrial and commercial applications. With its powerful 1/2 HP motor, it provides reliable performance. The open drip proof design allows it to be used in many environments. This electric motor would make a great addition for powering pumps, fans, conveyors, compressors, machine tools, and more.