Learn More
MSC Baldwin Air Filter Element, Axial Seal, Cellulose, 1 ea
Catalog No. 5000509622
Encompass
This round, axial seal outer air filter element is designed to protect internal combustion engines by preventing airborne contaminants from entering the combustion chamber. Constructed from high-quality cellulose and paper media, this filter ensures optimal engine performance and longevity in demanding applications.
  • Features securely affixed metal end caps with a raised gasket for a reliable seal to prevent leaks
  • Engineered for consistent filtration efficiency
  • Compatible with Kobelco excavators
  • Designed for use with inner air element PA4625
  • Overall dimensions: 374.7 mm (14.75 in.) height and 234.2 mm (9 7/32 in.) outside diameter
  • Includes attached and furnished gaskets for proper installation
Catalog No. 50-005-09622 Supplier MSC Supplier No. 55865257
This round, axial seal outer air filter element is designed to protect internal combustion engines by preventing airborne contaminants from entering the combustion chamber. Constructed from high-quality cellulose and paper media, this filter ensures optimal engine performance and longevity in demanding applications.
  • Features securely affixed metal end caps with a raised gasket for a reliable seal to prevent leaks
  • Engineered for consistent filtration efficiency
  • Compatible with Kobelco excavators
  • Designed for use with inner air element PA4625
  • Overall dimensions: 374.7 mm (14.75 in.) height and 234.2 mm (9 7/32 in.) outside diameter
  • Includes attached and furnished gaskets for proper installation