Learn More
MSC Baldwin Filters Air Filter, Outer, 204.8 mm Height, 1 ea
Catalog No. 5000508332
Encompass
This outer heavy-duty air filter element is designed to protect engines and mobile equipment by preventing airborne contaminants from reaching the combustion chamber. It features a robust design to support rigorous operating environments.
  • Axial seal design with securely affixed metal end caps
  • Integrated raised gasket ensures a proper seal and prevents leaks
  • Supports rigorous applications to ensure proper equipment function
Catalog No. 50-005-08332 Supplier MSC Supplier No. 55869002
This outer heavy-duty air filter element is designed to protect engines and mobile equipment by preventing airborne contaminants from reaching the combustion chamber. It features a robust design to support rigorous operating environments.
  • Axial seal design with securely affixed metal end caps
  • Integrated raised gasket ensures a proper seal and prevents leaks
  • Supports rigorous applications to ensure proper equipment function