Learn More
Medchemexpress LLC Outer membrane A/Omp 100ug
SDP
Catalog No. 5000196444
Encompass_Preferred
Outer membrane protein A (OmpA) ensures the structural integrity of the bacterial cell wall and cooperates with TolR to stabilize the peptidoglycan layer As a porin OmpA controls the permeability of small solutes Outer membrane protein A/OmpA Protein E coli (His-SUMO) is the recombinant E coli-derived Outer membrane protein A/OmpA protein expressed by E coli with N-SUMO N-6 His labeled tag
Catalog No. 50-001-96444 Supplier Medchemexpress LLC Supplier No. HYP71493100UG
Outer membrane protein A (OmpA) ensures the structural integrity of the bacterial cell wall and cooperates with TolR to stabilize the peptidoglycan layer As a porin OmpA controls the permeability of small solutes Outer membrane protein A/OmpA Protein E coli (His-SUMO) is the recombinant E coli-derived Outer membrane protein A/OmpA protein expressed by E coli with N-SUMO N-6 His labeled tag