Learn More
Waters Corp Outer Source Assembly, Xevo TQ-GC System, Source Heater, Focusing Optics
Catalog No. 502392241 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The outer source assembly is a crucial component of the ionization source within the Xevo TQ-GC system. Accessible from the instrument’s front, it integrates essential elements such as the source heater, PCB holder, and focusing optics.
  • Includes a source heater designed to raise temperature and prevent sample and matrix condensation.
  • Features focusing optics (lens stack) that focus the ion beam before it enters the analyzer.
  • Utilizes permanent magnets to spiral electrons into the inner source.
  • Designed for easy removal, enabling access and cleaning of its lenses.
  • Composed of a PCB holder, securing screw, 10-pin feed-through connector socket, and source block.
  • Contains two magnet holders, two magnet holder screws, a lens holder, and a PCB cable assembly.
Catalog No. 50-239-2241 Supplier Waters Corp Supplier No. 700011931
Add to Cart
Add to Cart
Request a Quote
The outer source assembly is a crucial component of the ionization source within the Xevo TQ-GC system. Accessible from the instrument’s front, it integrates essential elements such as the source heater, PCB holder, and focusing optics.
  • Includes a source heater designed to raise temperature and prevent sample and matrix condensation.
  • Features focusing optics (lens stack) that focus the ion beam before it enters the analyzer.
  • Utilizes permanent magnets to spiral electrons into the inner source.
  • Designed for easy removal, enabling access and cleaning of its lenses.
  • Composed of a PCB holder, securing screw, 10-pin feed-through connector socket, and source block.
  • Contains two magnet holders, two magnet holder screws, a lens holder, and a PCB cable assembly.