Learn More
ANALYTIK JENA US LLC Analytik Jena Quartz Glass Outer Tube, High Purity Quartz, V Suttle Torch Compatibility, Long Component Lifetime
Catalog No. NC1724209
Encompass
The Analytik Jena Quartz Glass Outer Tube is essential for maintaining plasma integrity within the PlasmaQuant 9100 ICP-OES series. Its high-purity quartz glass construction ensures efficient sample atomization and excitation. This robust outer tube minimizes maintenance needs and reduces issues related to washout and sample deposition, thereby enhancing analytical precision and accuracy, making it a critical component for reliable elemental analysis.
Catalog No. NC1724209 Supplier ANALYTIK JENA US LLC Supplier No. 41813410402
The Analytik Jena Quartz Glass Outer Tube is essential for maintaining plasma integrity within the PlasmaQuant 9100 ICP-OES series. Its high-purity quartz glass construction ensures efficient sample atomization and excitation. This robust outer tube minimizes maintenance needs and reduces issues related to washout and sample deposition, thereby enhancing analytical precision and accuracy, making it a critical component for reliable elemental analysis.