Learn More
Saf T Pak Inc STP-211 Outer Box for STP-210, Refurbishment (UN 3373, Exempt), Outer Box 25/Case
SDP
Catalog No. NC1007938 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
System Components:Outer Box OD 9 x 4 x 4", pre-printed with: Biological Substance, Category B and UN 3373 Marking (Category B Side), Exempt Human/ Animal Specimen Checkbox Markings (Exempt side)
Catalog No. NC1007938 Supplier Saf T Pak Inc Supplier No. STP211
System Components:Outer Box OD 9 x 4 x 4", pre-printed with: Biological Substance, Category B and UN 3373 Marking (Category B Side), Exempt Human/ Animal Specimen Checkbox Markings (Exempt side)