Learn More
Waters Corp Waters Outlet Check Valve Rebuild Kit, 225μL
Catalog No. 50793537 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit allows users to assemble and replace valves themselves, offering significant cost savings. While pre-assembled ball and seat outlet check valves are available, this rebuild kit provides a self-maintenance option.
  • Allows self-assembly and replacement of valves for significant cost savings.
  • Provides the option of self-maintenance.
  • Contains the ball and seat cartridge, gaskets, and comprehensive instructions.
Catalog No. 50-793-537 Supplier Waters Corp Supplier No. WAT026014
This kit allows users to assemble and replace valves themselves, offering significant cost savings. While pre-assembled ball and seat outlet check valves are available, this rebuild kit provides a self-maintenance option.
  • Allows self-assembly and replacement of valves for significant cost savings.
  • Provides the option of self-maintenance.
  • Contains the ball and seat cartridge, gaskets, and comprehensive instructions.