Learn More
CMP SCIENTIFIC CORP Outlet tapered separation capillary, CC1 coated
Catalog No. NC2205089
Encompass
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 70 cm, CC1 coated
This item is not returnable. View return policy
Catalog No. NC2205089 Supplier CMP SCIENTIFIC CORP Supplier No. ESCCC1360/1505070B1
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 70 cm, CC1 coated