Learn More
Grainger Fill-Rite Flange Kit, Straight 1 in. Discharge, 700V/710V Series, 1 ea.
Catalog No. 502782511
Encompass_Preferred
This outlet flange kit provides a replacement straight 1 in. discharge flange for specific fuel transfer pumps. Designed for seamless integration, this kit includes the necessary components to maintain secure connections in your pump system.
  • Straight 1 in. discharge flange with 1 in. NPT threads
  • Includes discharge flange, flange bolts, O-ring, and button head cap screw
  • Compatible with FR700V and FR710V series pumps
  • Constructed from cast iron, copper, fluorocarbon, sintered iron, and steel
  • Chemical compatibility includes biodiesel, diesel, E15, gasoline, kerosene, and mineral spirits
Catalog No. 50-278-2511 Supplier Grainger Supplier No. 19NL50
This outlet flange kit provides a replacement straight 1 in. discharge flange for specific fuel transfer pumps. Designed for seamless integration, this kit includes the necessary components to maintain secure connections in your pump system.
  • Straight 1 in. discharge flange with 1 in. NPT threads
  • Includes discharge flange, flange bolts, O-ring, and button head cap screw
  • Compatible with FR700V and FR710V series pumps
  • Constructed from cast iron, copper, fluorocarbon, sintered iron, and steel
  • Chemical compatibility includes biodiesel, diesel, E15, gasoline, kerosene, and mineral spirits