Learn More
Chemglass Life Sciences ChemRxnHub Manifold System to Include: 1ea CG-1969-X-12 (Outlet Manifold Assembly Complete) and 1ea CG-1969-X-14 (Inlet Manifold Assembly Complete)
SDP
Catalog No. 50143599
Encompass_Preferred
ChemRxnHub Manifold System to Include: 1ea CG-1969-X-12 (Outlet Manifold Assembly Complete) and 1ea CG-1969-X-14 (Inlet Manifold Assembly Complete). The Chemglass manifold system for use with CG-1949-X ChemglassRxnHub™ frame system allows for easy and safe connection of your circulator to our glass process reactors. Manifold helps alleviate excessive mechanical forces placed on the glass vessel inlet/outlet. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on S.S. Tool-Free™ fitting on one end and M16 Female fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately. PLEASE NOTE: TEMPERATURE LIMITATIONS ON MANIFOLD IS -60 TO 200°C. Specifications:Circulator Size End Fitting: M16 For Use with Reactor Capacity: 300mL to 5L Reactor Jacket Fitting: Tool-Free™ Fits Support Frame: CG-1949-X S.S. Pipe OD: 3/4"
This item is not returnable. View return policy
Catalog No. 50-143-599 Supplier Chemglass Life Sciences Supplier No. CG1969X10
ChemRxnHub Manifold System to Include: 1ea CG-1969-X-12 (Outlet Manifold Assembly Complete) and 1ea CG-1969-X-14 (Inlet Manifold Assembly Complete). The Chemglass manifold system for use with CG-1949-X ChemglassRxnHub™ frame system allows for easy and safe connection of your circulator to our glass process reactors. Manifold helps alleviate excessive mechanical forces placed on the glass vessel inlet/outlet. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on S.S. Tool-Free™ fitting on one end and M16 Female fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately. PLEASE NOTE: TEMPERATURE LIMITATIONS ON MANIFOLD IS -60 TO 200°C. Specifications:Circulator Size End Fitting: M16 For Use with Reactor Capacity: 300mL to 5L Reactor Jacket Fitting: Tool-Free™ Fits Support Frame: CG-1949-X S.S. Pipe OD: 3/4"