Learn More
AB Sciex LLC OUTLET PLATE PACK
Catalog No. NC2157551
Encompass
BioPhase 8800 Outlet plate pack includes 20 outlet plates. Plates include 96 wells and are designed to ANSI standard configuration for compatibility with automated 96-well plate handlers. Plate design enables use of 8-capillary BioPhase cartridges.
This item is not returnable. View return policy
Catalog No. NC2157551 Supplier AB Sciex LLC Supplier No. 5080315
BioPhase 8800 Outlet plate pack includes 20 outlet plates. Plates include 96 wells and are designed to ANSI standard configuration for compatibility with automated 96-well plate handlers. Plate design enables use of 8-capillary BioPhase cartridges.