Learn More
CMP SCIENTIFIC CORP Outlet tapered separation capillary, PS1 coated
Catalog No. NC2318826
Encompass
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 100 cm, PS1 coated
Catalog No. NC2318826 Supplier CMP SCIENTIFIC CORP Supplier No. ESCPS1360/15050100B
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 100 cm, PS1 coated