Learn More
CMP SCIENTIFIC CORP Outlet tapered separation capillary, PS2 coated
Catalog No. NC2318828
Encompass
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 100 cm, PS2 coated
Catalog No. NC2318828 Supplier CMP SCIENTIFIC CORP Supplier No. ESCPS2360/15050100B
Outlet tapered separation capillary (1 per box): 360/150 µm OD, 50 µm ID, 100 cm, PS2 coated