Learn More
Agilent Technologies Outlet Valve, 600 Bar, For HPLC Pumps, 1 Ea.
Catalog No. NC2080628
Encompass
This outlet valve is a critical component designed for optimal performance in liquid chromatography applications. Engineered to ensure reliable flow regulation within the pump assembly, it supports consistent system pressure and precise operation.
  • Designed for integration with 1100, 1200, and 1260 Infinity series pumps
  • Compatible with analytical liquid chromatography systems
  • Features a maximum operating pressure of 600 bar
  • Includes an integrated seal for improved functionality
Catalog No. NC2080628 Supplier Agilent Technologies Supplier No. G131260067
This outlet valve is a critical component designed for optimal performance in liquid chromatography applications. Engineered to ensure reliable flow regulation within the pump assembly, it supports consistent system pressure and precise operation.
  • Designed for integration with 1100, 1200, and 1260 Infinity series pumps
  • Compatible with analytical liquid chromatography systems
  • Features a maximum operating pressure of 600 bar
  • Includes an integrated seal for improved functionality