Learn More
Grainger Wheeler-Rex Discharge Hose, High Pressure, 1/4 in. ID, 10 ft., 3,000 psi, 1 ea.
Catalog No. 502785217
Encompass_Preferred
This high pressure discharge hose is designed for use with hydrostatic test pumps. The 10 ft. assembly features a durable construction with a 1/4 in. inside diameter, suitable for demanding testing environments.
  • Operating pressure of 3,000 psi
  • 1/4 in. inside diameter
  • 10 ft. length
  • Features 9/16-18 JICF female swivel x 3/8 MNPT connection ends
  • Compatible with hydrostatic test pumps
Catalog No. 50-278-5217 Supplier Grainger Supplier No. 19C695
This high pressure discharge hose is designed for use with hydrostatic test pumps. The 10 ft. assembly features a durable construction with a 1/4 in. inside diameter, suitable for demanding testing environments.
  • Operating pressure of 3,000 psi
  • 1/4 in. inside diameter
  • 10 ft. length
  • Features 9/16-18 JICF female swivel x 3/8 MNPT connection ends
  • Compatible with hydrostatic test pumps