Learn More
GVS FILTER TECHNOLOGY INC Elipse OV/AG/P100 pair of filters for SPR472/473
Catalog No. NC1600858
Encompass_Preferred
This item has a minimum qty of 8 per supplier requirements.
Replaceable OV/AG P100 filters for the SPR472/473. Large surface area for better communication and easy breathability. Activated carbon is good to protect against Organic Vapors and Acid Gases as well as dust mists and fumes. Comes with two OV/AG P100 filters.
Catalog No. NC1600858 Supplier GVS FILTER TECHNOLOGY INC Supplier No. SPR474
This item has a minimum qty of 8 per supplier requirements.
Replaceable OV/AG P100 filters for the SPR472/473. Large surface area for better communication and easy breathability. Activated carbon is good to protect against Organic Vapors and Acid Gases as well as dust mists and fumes. Comes with two OV/AG P100 filters.