Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Comfort-Air™ Series 300 Half-Face Respirators with OV/N95 Filter Assembly
SDP

Catalog No. 19222457 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
Small/Medium, Display Bag Packaged
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-222-457 Small/Medium, Display Bag Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-457 Supplier Dentec Safety™ Supplier No. 3SM20DN5ZAM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermoplastic respirators with organic vapor N95 filter assembly are suitable for use with lacquer, enamel paints, pesticides, welding, dusts and non-oil mist applications.

  • Extended field of vision
  • Large inhalation valves for maximum inhalation
  • Strategically positioned exhalation valve under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Exhalation valve diaphragm has channels that act like a parachute to traps exhaled air and lift the diaphragm off the valve seat with very little effort
  • Rolled sealing flange provides comfort and improves fitting capabilities. Does not require user to adjust straps excessively tight
  • Textured sealing area reduces risk of the mask slipping on the face
  • Comfortable on a wide variety of facial sizes
  • Enhanced harness assembly features:
    • Metal tabs at end to prevent nylon strap fraying and allows for adjustment when wearing gloves
    • Wide nylon strap maintains elasticity longer
    • 4-point swiveling adjustments
    • Cradle suspension fits comfortably under hard hats, grinding and welding helmets
  • 3-position adjustment allows users to adjust cartridge to three different positions

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Gray
Filter Type N95
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small/Medium
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Application) Lacquer, enamel paints, pesticides, welding, dusts
For Use With (Equipment) Cartridges/Diskits
Material Headstrap Nylon
Model 3-SM-20DN5ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 17.78 cm
Height (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.