Learn More
Foxx Life Sciences EZBio MTO Single Use (SUT) Media Bottle Assembly, 250mL, PC, 38-430 VersaCap with Over molded Platinum-Cured Silicone Tubing, Vented Port with Diptube and Plug, Aseptic Transfer System, Sterile, 10/Case
GREENER_CHOICE
Catalog No. 5000575200
Encompass_Preferred
EZBio Media Bottle Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use closed system designed for aseptic fluid transfer, eliminating in-house assembly and validation to save time and reduce risk. It removes the need to source or qualify individual components.
Available in PETG or polycarbonate (PC) square bottles (60 mL to 2 L), each assembly includes a preassembled VersaCap with molded silicone adapter, 1/8” tubing, male and female connectors, dip tube, 0.22 µm vent filter, and pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, these bioprocess containers (BPC) offer a cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom
Catalog No. 50-005-75200 Supplier Foxx Life Sciences Supplier No. 438E004FLS
EZBio Media Bottle Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use closed system designed for aseptic fluid transfer, eliminating in-house assembly and validation to save time and reduce risk. It removes the need to source or qualify individual components.
Available in PETG or polycarbonate (PC) square bottles (60 mL to 2 L), each assembly includes a preassembled VersaCap with molded silicone adapter, 1/8” tubing, male and female connectors, dip tube, 0.22 µm vent filter, and pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, these bioprocess containers (BPC) offer a cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom