Learn More
Foxx Life Sciences EZBio Single Use (SUT) Media Bottle Cap Assembly, 1L, 38-430 VersaCap with Overmolded Platinum Cured Silicone Tubing with Luer, Vented Port with Dip tube, Aseptic Transfer Cap, Sterile, 20/CS
Catalog No. 5000575146
Encompass_Preferred
EZBio Transfer Cap Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use bioprocess container (BPC) Cap assembly for aseptic transfer system that eliminates in-house assembly and validation, saving time and reducing risk. It removes the need to source or qualify individual components.
Available for square bottles (125mL to 2L) in PETG or polycarbonate (PC), each assembly features a preassembled VersaCap with molded silicone adapter, 1/8” tubing, connectors, dip tube, 0.22 µm vent filter, transfer cap, and ratchet-style pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, it provides a cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom
Catalog No. 50-005-75146 Supplier Foxx Life Sciences Supplier No. 430E217FLS
EZBio Transfer Cap Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use bioprocess container (BPC) Cap assembly for aseptic transfer system that eliminates in-house assembly and validation, saving time and reducing risk. It removes the need to source or qualify individual components.
Available for square bottles (125mL to 2L) in PETG or polycarbonate (PC), each assembly features a preassembled VersaCap with molded silicone adapter, 1/8” tubing, connectors, dip tube, 0.22 µm vent filter, transfer cap, and ratchet-style pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, it provides a cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom