Learn More
Foxx Life Sciences EZBiopure TR Single Use (SUT) Media Bottle Assembly, 125mL, PETG, GL45 wide mouth VersaCap with Over molded Platinum Cured Silicone Tubing, Vented Port with Dip Tube & MicroCNX, Aseptic Transfer Assembly, Sterile, 10/CS
GREENER_CHOICE
Catalog No. 5000575147
Encompass_Preferred
The EZBiopure TR Wide Mouth Media Bottle Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use closed system bioprocess container designed for aseptic fluid transfer, eliminating in-house assembly and validation to save time and reduce risk. It removes the need to source or qualify individual components.
Available in 125 mL to 1L round bottles, in PETG or Polysulfone (PS), each assembly includes a preassembled VersaCap with molded silicone adapter, 1/8” tubing, connectors, dip tube, 0.22 µm vent filter, and ratchet-style pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, these assemblies provide a convenient, cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom
Catalog No. 50-005-75147 Supplier Foxx Life Sciences Supplier No. 52744E5FLS
The EZBiopure TR Wide Mouth Media Bottle Assembly with Molded Silicone Cap Adapter is a ready-to-use, single-use closed system bioprocess container designed for aseptic fluid transfer, eliminating in-house assembly and validation to save time and reduce risk. It removes the need to source or qualify individual components.
Available in 125 mL to 1L round bottles, in PETG or Polysulfone (PS), each assembly includes a preassembled VersaCap with molded silicone adapter, 1/8” tubing, connectors, dip tube, 0.22 µm vent filter, and ratchet-style pinch clamp.
The one-piece design minimizes leaks, contamination, and product loss. Sterile, gamma-irradiated, and individually packaged, these assemblies provide a convenient, cost-effective alternative to stainless-steel systems requiring cleaning and validation.
Features:
Joint-free, leak-proof molded silicone cap adapter
Low-extractables tubing
Manufactured in ISO Class 7 cleanroom