Learn More
Jeio Tech Inc Jeio Tech™ OV4-30, 28L Vacuum Oven (120V, 60Hz), Drying for Heat-Sensitive Materials, PID Temperature Control, Uniform Heating
SDP
Catalog No. NC3662716
Encompass_Preferred
Jeio Tech OV4-30S Vacuum Oven delivers precise, gentle drying for heat-sensitive, easily oxidized, and powdered materials. By reducing the boiling point under vacuum, the OV4 enables efficient drying while minimizing thermal degradation and oxidation. Built with a corrosion-resistant chamber and intuitive controls, it ensures reliable and repeatable performance. The PID control system delivers excellent stability (±0.2°C fluctuation) and uniformity (±1.5°C variation) across a temperature range of Ambient +15°C to 250°C. Standard aluminum shelves enhance heat distribution and durability. Additional features include auto-tuning, 3-point calibration, 3.5” LCD touchscreen, and safety protections. GMP-ready options include BMS port (4-20mA) and 21 CFR Part 11 compliant software. Inner chamber: W 11.9 × D 12 × H 11.9 inches.
Catalog No. NC3662716 Supplier Jeio Tech Inc Supplier No. AAH13336U
Jeio Tech OV4-30S Vacuum Oven delivers precise, gentle drying for heat-sensitive, easily oxidized, and powdered materials. By reducing the boiling point under vacuum, the OV4 enables efficient drying while minimizing thermal degradation and oxidation. Built with a corrosion-resistant chamber and intuitive controls, it ensures reliable and repeatable performance. The PID control system delivers excellent stability (±0.2°C fluctuation) and uniformity (±1.5°C variation) across a temperature range of Ambient +15°C to 250°C. Standard aluminum shelves enhance heat distribution and durability. Additional features include auto-tuning, 3-point calibration, 3.5” LCD touchscreen, and safety protections. GMP-ready options include BMS port (4-20mA) and 21 CFR Part 11 compliant software. Inner chamber: W 11.9 × D 12 × H 11.9 inches.