Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OVA (257-264), scrambled | 1213245-88-5 | 98.29% | 963.13 | 1 MG
SDP

Catalog No. 5000398045
Encompass_Preferred

OVA (257-264), scrambled is a biologically active peptide. It serves as an out-of-order control peptide for HY-P1489 and is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Sequence: Phe-Ile-Leu-Lys-Ser-Ile-Asn-Glu
  • Sequence shortening: FILKSINE
  • Soluble in DMSO

Catalog No. 50-003-98045 Supplier Medchemexpress LLC Supplier No. HYP54001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OVA (257-264), scrambled is a biologically active peptide. It serves as an out-of-order control peptide for HY-P1489 and is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Sequence: Phe-Ile-Leu-Lys-Ser-Ile-Asn-Glu
  • Sequence shortening: FILKSINE
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.