Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OVA (257-264), scrambled | 1213245-88-5 | 98.3% | 963.13 | 10 MG
SDP

Catalog No. 5000399129
Encompass_Preferred

OVA (257-264), scrambled is a biological active peptide that serves as an out-of-order control peptide of HY-P1489.

  • Biological active peptide
  • Out-of-order control peptide of HY-P1489
  • Purity of 98.29%
  • Appears as a white to off-white solid
  • Storage as powder: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture and light)
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-99129 Supplier Medchemexpress LLC Supplier No. HYP540010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OVA (257-264), scrambled is a biological active peptide that serves as an out-of-order control peptide of HY-P1489.

  • Biological active peptide
  • Out-of-order control peptide of HY-P1489
  • Purity of 98.29%
  • Appears as a white to off-white solid
  • Storage as powder: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture and light)
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.