Learn More
CPC Scientific H-Ile-Ser-Gln-Ala-Val-His-Ala-Ala-His-Ala-Glu-Ile-Asn-Glu-Ala-Gly-Arg-OH 1MG
SDP
Supplier:  CPC Scientific MISC011B
Encompass
An H-2b-restricted OVA class II epitope used in studying T-cell activation and allergic diseases.SEQUENCE: H-Ile-Ser-Gln-Ala-Val-His-Ala-Ala-His-Ala-Glu-Ile-Asn-Glu-Ala-Gly-Arg-OHONE-LETTER SEQUENCE: ISQAVHAAHAEINEAGRMOLECULAR FORMULA: C74H120N26O25MOLECULAR WEIGHT: 1773.93STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [92915-79-2]SYNONYMS: OVA (323-339) (chicken, japanese quail), Ovalbumin (323-339) (chicken, japanese quail)RESEARCH AREA: MiscellaneousREFERENCES:K. M. Murphy, Science 250, 1760Oran, A. and H. Robinson, J. Immunol. 171, 1999 (2003)Palmiter, RD. et al. Proc. Natl. Acad. Sci. 75, 94 (1978)
SKU(s): MISC-011A, MISC-011B
Catalog No. 50-210-9734